missing translation for 'onlineSavingsMsg'
Learn More

Acetaldehyde ammonia trimer, 98%

Product Code. 15498949
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15498949 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15498949 Supplier Thermo Scientific Alfa Aesar Supplier No. L12265.18
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 58052-80-5 | C6H18N3 | 132.23 g/mol

Acetaldehyde ammonia trimer is used as scavengers for sulfhydryl compounds in natural gas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetaldehyde ammonia trimer is used as scavengers for sulfhydryl compounds in natural gas.

Solubility
Soluble in water.

Notes
Desiccate at 4°C. Store away from air, moisture and oxidizing agents
TRUSTED_SUSTAINABILITY

Specifications

CAS 58052-80-5
Melting Point 93°C to 96°C
Density 0.852
Boiling Point 110°C (decomposition)
Molecular Formula C6H18N3
MDL Number MFCD00149559
Quantity 50 g
UN Number UN1841
Beilstein 3910586
Sensitivity Hygroscopic
Merck Index 14,40
Synonym 2,4,6-trimethyl-1,3,5-triazinane trihydrate, acetaldehyde ammonia trimer, hexahydro-2,4,6-trimethyl-1,3,5-triazine trihydrate, 1,3,5-triazine, hexahydro-2,4,6-trimethyl-, trihydrate, acetaldehyde ammonia trimer trihydrate, 2,4,6-trimethyl-1,3,5-triazaperhydroine, hydrate, hydrate, hydrate, acetaldehydeammoniatrimer, acmc-1bm0n, acetaldehyde-ammonia trimer trihydrate, acetaldehyde ammonia trimer nt
Solubility Information Soluble in water.
InChI Key MZSSRMMSFLVKPK-UHFFFAOYSA-Q
SMILES CC1[NH2+]C(C)[NH2+]C(C)[NH2+]1
IUPAC Name 2,4,6-trimethyl-1,3,5-triazinane;trihydrate
Molecular Weight (g/mol) 132.23
PubChem CID 2723814
Formula Weight 183.25
Percent Purity 98%
Chemical Name or Material Acetaldehyde ammonia trimer
Show More Show Less
compliance-icons
Product Identifier
  • Acetaldehyde ammonia trimer
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contain: Acetaldehyde-ammonia

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.