missing translation for 'onlineSavingsMsg'
Learn More

Acetaldehyde, 99.5%, extra pure

Product Code. 10423215 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
10mL
2.5 L
250mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10423215 2.5 L Glass bottle 2.5 L
10705041 10 mL Glass bottle 10mL
10061530 250 mL Glass bottle 250mL
10051530 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10423215 Supplier Thermo Scientific Acros Supplier No. 149510025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 75-07-0 | C2H4O | 44.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-07-0
Molecular Formula C2H4O
Molecular Weight (g/mol) 44.04
MDL Number MFCD00006991
InChI Key IKHGUXGNUITLKF-UHFFFAOYSA-N
Synonym ethanal, acetic aldehyde, ethyl aldehyde, acetaldehyd, acetylaldehyde, aldehyde, acetic ethanol, aldeide acetica, octowy aldehyd, aldehyde acetique
PubChem CID 177
ChEBI CHEBI:15343
IUPAC Name acetaldehyde
SMILES CC=O

Specifications

CAS Min % 99.4
CAS Max % 100.0
Melting Point -123.0°C
Color Colorless
Density 0.7850g/mL
Boiling Point 21.0°C
Flash Point −27°C
Infrared Spectrum Authentic
Assay Percent Range 99.4% min. (GC)
Odor Pungent
Packaging Glass bottle
Linear Formula CH3CHO
Refractive Index 1.3310 to 1.3330 (20°C, 589nm)
Quantity 2.5 L
Beilstein 01, 594
Fieser 01,3
Merck Index 15, 40
Specific Gravity 0.785
Solubility Information Solubility in water: > 500g/L (20°C). Other solubilities: soluble in ethanol and acetone
Formula Weight 44.04
Percent Purity 99.5%
Grade Extra Pure
Physical Form Volatile Liquid
Chemical Name or Material Acetaldehyde, Extra Pure
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.