missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetaldehyde 2,4-Dinitrophenylhydrazone, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Acetaldehyde-2,4-dinitrophenylhydrazone (DNPH) |
| CAS | 1019-57-4 |
| Molecular Formula | C8 H8 N4 O4 |
| Quantity | 10 mg |
| Synonym | Acetaldehyde, (2,4-dinitrophenyl)hydrazone (6CI,8CI,9CI), Ethanal-2,4-dinitrophenylhydrazone, NSC 403216 |
| InChI Formula | InChI=1S/C8H8N4O4/c1-2-9-10-7-4-3-6(11(13)14)5-8(7)12(15)16/h2-5,10H,1H3/b9-2+ |
| SMILES | C\C=N\Nc1ccc(cc1[N+](=O)[O-])[N+](=O)[O-] |
| IUPAC Name | N-[(E)-ethylideneamino]-2,4-dinitroaniline |
| Molecular Weight (g/mol) | 224.17 |
| Formula Weight | 224.0546 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction