missing translation for 'onlineSavingsMsg'
Learn More

Acetaldehyde-2,4-dinitrophenylhydrazone 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30314333
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30314333 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30314333 Supplier Dr. Ehrenstorfer Supplier No. DREV10011300AL100
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Especificaciones

Chemical Name or Material Acetaldehyde-2,4-dinitrophenylhydrazone (DNPH)
CAS 1019-57-4
Analyte or Component Names Acetaldehyde-2,4-dinitrophenylhydrazone (DNPH)
Shipping Condition Room Temperature
Molecular Formula C8 H8 N4 O4
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Acetaldehyde, (2,4-dinitrophenyl)hydrazone (6CI,8CI,9CI), Ethanal-2,4-dinitrophenylhydrazone, NSC 403216
InChI Formula InChI=1S/C8H8N4O4/c1-2-9-10-7-4-3-6(11(13)14)5-8(7)12(15)16/h2-5,10H,1H3/b9-2+
SMILES C\C=N\Nc1ccc(cc1[N+](=O)[O-])[N+](=O)[O-]
IUPAC Name N-[(E)-ethylideneamino]-2,4-dinitroaniline
Solvent or Matrix Acetonitrile
Industry Type Food Testing
Molecular Weight (g/mol) 224.17
Quantity 5 mL
Formula Weight 224.0546 g/mol
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.