missing translation for 'onlineSavingsMsg'
Learn More

Acetal, TRC

Product Code. 17960505
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17960505 10 g 10g
17931035 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17960505 Supplier Toronto Research Chemicals Supplier No. TRCA13259010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-57-7
Molecular Formula C6 H14 O2
Molecular Weight (g/mol) 118.17
Synonym Acetaldehyde, diethyl acetal (8CI), 1,1-Diethoxyethane, Acetal, Acetaldehyde ethyl acetal, Diethyl acetal, Ethanal diethyl acetal, Ethylidene diethyl ether, NSC 7624
IUPAC Name 1,1-diethoxyethane
SMILES CCOC(C)OCC

Specifications

Quantity 10 g
InChI Formula InChI=1S/C6H14O2/c1-4-7-6(3)8-5-2/h6H,4-5H2,1-3H3
Formula Weight 118.0994
Percent Purity >95%
Chemical Name or Material Acetaldehyde diethylacetal
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.