missing translation for 'onlineSavingsMsg'
Learn More

Acetal, 99%

Product Code. 10706321 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10706321 5 mL Glass bottle 5mL
10687432 250 mL Glass bottle 250mL
10588650 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10706321 Supplier Thermo Scientific Acros Supplier No. 102220050
  • ONLINE EXCLUSIVE PRICE
    £20.00
    £18.65 / 5mL
    Save £1.35
    6% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 105-57-7 | C6H14O2 | 118.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-57-7
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.18
MDL Number MFCD00009243
InChI Key DHKHKXVYLBGOIT-UHFFFAOYSA-N
Synonym acetal, acetaldehyde diethyl acetal, diethyl acetal, ethane, 1,1-diethoxy, diaethylacetal, acetale, ethylidene diethyl ether, acetal diethylique, diethylacetal, 1,1-dietossietano
PubChem CID 7765
IUPAC Name 1,1-diethoxyethane
SMILES CCOC(C)OCC

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -100.0°C
Color Colorless
Density 0.8250g/mL
Boiling Point 103.0°C
Flash Point −21°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH(OC2H5)2
Refractive Index 1.3790 to 1.3850
Quantity 5 mL
Beilstein 01, 603
Merck Index 15, 39
Specific Gravity 0.825
Solubility Information Solubility in water: 46g/L (25°C). Other solubilities: miscible with alcohol,ether,soluble in heptane,methylcyclohexane,,ethyl acetate,propyl-,isopropyl-,butyl-,,isobutyl alcohol
Formula Weight 118.18
Percent Purity 99%
Grade Pure
Physical Form Liquid
Chemical Name or Material Acetal, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetal
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.