missing translation for 'onlineSavingsMsg'
Learn More

Acesulfame Aspartame Salt, TRC

Product Code. 30180221
Change view
Click to view available options
Quantity:
200 mg
500 mg
1 g
Unit Size:
1g
200mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30180221 1 g 1g
30180048 200 mg 200mg
30180365 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30180221 Supplier Toronto Research Chemicals Supplier No. TRCA1325201G
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106372-55-8
Synonym Acesulfame Aspartame Salt∣Aspartame Acesulfame

Specifications

Format Neat
Molecular Weight (g/mol) 457.45
Product Type Impurity
Quantity 1 g
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.