missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals ACES, 99%
Click to view available options
Quantity:
25 g
100 g
500 g
Description
ACES is used as a Good's buffer is an ideal separator in the resolution of protein systems, a zwitterionic compound used in biochemical and biological research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
ACES is used as a Good′s buffer is an ideal separator in the resolution of protein systems, a zwitterionic compound used in biochemical and biological research.
Solubility
Soluble in water(18g/L).
Notes
Air Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
ACES is used as a Good′s buffer is an ideal separator in the resolution of protein systems, a zwitterionic compound used in biochemical and biological research.
Solubility
Soluble in water(18g/L).
Notes
Air Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Spécification
Melting Point | ∼275°C (decomposition) |
Quantity | 25 g |
Beilstein | 2253770 |
Merck Index | 14,36 |
Solubility Information | Soluble in water(18g/L). |
Formula Weight | 182.2 |
Percent Purity | 99% |
Chemical Name or Material | ACES |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'