missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACER3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584834
This item is not returnable.
View return policy
Description
Immunogen sequence: DREGYWGPTT STLDWCEENY SVTWYIAEFW NTVSNLIMII PPMFGAVQSV RDGLEKR Highest antigen sequence indentity to the following orthologs: Mouse - 82%, Rat - 82%.
ACER3 is a protein coding gene. Endoplasmic reticulum and Golgi ceramidase that catalyzes the hydrolysis of unsaturated long-chain C18:1-, C20:1- and C20:4-ceramides, dihydroceramides and phytoceramides into sphingoid bases like sphingosine and free fatty acids at alkaline pH. Ceramides, sphingosine, and its phosphorylated form sphingosine-1-phosphate are bioactive lipids that mediate cellular signaling pathways regulating several biological processes including cell proliferation, apoptosis and differentiation. Controls the generation of sphingosine in erythrocytes, and thereby sphingosine-1-phosphate in plasma. Through the regulation of ceramides and sphingosine-1-phosphate homeostasis in the brain may play a role in neurons survival and function. By regulating the levels of proinflammatory ceramides in immune cells and tissues, may modulate the inflammatory response.
Specifications
| ACER3 | |
| Polyclonal | |
| Unconjugated | |
| ACER3 | |
| 1110057L18Rik; 5430429L08Rik; Acer3; alkaline CDase 3; alkaline ceramidase 3; Alkaline dihydroceramidase SB89; alkaline phytoceramidase; AlkCDase 3; aPHC; AV015045; Phca; phytoceramidase, alkaline; RGD1561254 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55331 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NUN7 | |
| ACER3 | |
| Recombinant protein corresponding to Human ACER3. Recombinant protein control fragment (Product #RP-108561). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction