Få mere at vide
Invitrogen™ ACER2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114249
Beskrivelse
Immunogen sequence: DELAVLWVLM CALAMWFPRR YLPKIFRNDR GRFKVVVSVL SAVTTCLAFV For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.
The sphingolipid metabolite sphingosine-1-phosphate promotes cell proliferation and survival, whereas its precursor, sphingosine, has the opposite effect. The ceramidase ACER2 hydrolyzes very long chain ceramides to generate sphingosine.
Tekniske data
| ACER2 | |
| Polyclonal | |
| Unconjugated | |
| ACER2 | |
| 2410116I05Rik; ACER2; Acylsphingosine deacylase 3-like; alkaline CDase 2; alkaline ceramidase 2; AlkCDase 2; ALKCDase2; Asah3l; cancer related gene-liver 1 (CRG-L1); cancer-related gene liver 1 protein; ceramide hydrolase; CRG-L1; haCER2; maCER2; N-acylsphingosine amidohydrolase 3-like; PP11646 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 340485 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q5QJU3 | |
| ACER2 | |
| Synthetic peptide directed towards the middle region of Human ACER2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.