missing translation for 'onlineSavingsMsg'
Learn More

Acepromethazine, TRC

Product Code. 30008535
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008535 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30008535 Supplier Toronto Research Chemicals Supplier No. TRCA13091010MG
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 13461-01-3
Molecular Formula C19H22N2OS
Quantity 10 mg
Synonym 1-[10-[2-(Dimethylamino)propyl]-10H-phenothiazin-2-yl]ethanone, 10-[2-(Dimethylamino)propyl]phenothiazin-2-yl Methyl Ketone, 1664CB, Aceprometazine
InChI Formula InChI=1S/C19H22N2OS/c1-13(20(3)4)12-21-16-7-5-6-8-18(16)23-19-10-9-15(14(2)22)11-17(19)21/h5-11,13H,12H2,1-4H3
SMILES CC(CN1c2ccccc2Sc3ccc(cc13)C(=O)C)N(C)C
IUPAC Name 1-[10-[2-(dimethylamino)propyl]phenothiazin-2-yl]ethanone
Molecular Weight (g/mol) 326.46
Chemical Name or Material Acepromethazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.