missing translation for 'onlineSavingsMsg'
Learn More

Acepromazine Maleate, Mikromol™

Product Code. 17932736
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17932736 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17932736 Supplier Mikromol Supplier No. MM0927.00
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Acepromazine Maleate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acepromazine maleate salt
CAS 3598-37-6
Analyte or Component Names Acepromazine maleate salt
Shipping Condition Room Temperature
Molecular Formula C19 H22 N2 O S . C4 H4 O4
Certifications/Compliance ISO 17034
Synonym Ethanone, 1-[10-[3-(dimethylamino)propyl]-10H-phenothiazin-2-yl]-, (Z)-2-butenedioate (1:1), Ketone, 10-[3-(dimethylamino)propyl]phenothiazin-2-yl methyl, maleate (1:1) (8CI), Ketone, 10-[3-(dimethylamino)propyl]phenothiazin-2-yl methyl, maleate (6CI,7CI), Maleic acid, compd. with 10-(3-dimethylaminopropyl)phenothiazin-2-yl methyl ketone (6CI), Acepran, Acepromazine maleate, Acepromazine monomaleate, Acetopromazine maleate salt, Acetylpromazine maleate, Anatran, Atravet, Calmivet, Calmo Neosan, NSC 264522, Notensil, Notenzil, Plegicil, Sedalin, Soprontin
InChI Formula InChI=1S/C19H22N2OS.C4H4O4/c1-14(22)15-9-10-19-17(13-15)21(12-6-11-20(2)3)16-7-4-5-8-18(16)23-19;5-3(6)1-2-4(7)8/h4-5,7-10,13H,6,11-12H2,1-3H3;1-2H,(H,5,6)(H,7,8)/b;2-1-
SMILES CN(C)CCCN1c2ccccc2Sc3ccc(cc13)C(=O)C.OC(=O)\C=C/C(=O)O
IUPAC Name (Z)-but-2-enedioic acid;1-[10-[3-(dimethylamino)propyl]phenothiazin-2-yl]ethanone
Molecular Weight (g/mol) 442.53
Quantity 250 mg
Formula Weight 442.1562 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.