missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Acepromazine maleate, 98%

Product Code. 15892524 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15892524 1 g 1g
15802534 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15892524 Supplier Thermo Scientific Chemicals Supplier No. 462590010
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 3598-37-6 | C23H26N2O5S | 442.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3598-37-6
Molecular Formula C23H26N2O5S
Molecular Weight (g/mol) 442.53
InChI Key FQRHOOHLUYHMGG-WLHGVMLRSA-N
IUPAC Name (2E)-but-2-enedioic acid; 1-{10-[3-(dimethylamino)propyl]-10H-phenothiazin-2-yl}ethan-1-one
SMILES OC(=O)\C=C\C(O)=O.CN(C)CCCN1C2=CC=CC=C2SC2=CC=C(C=C12)C(C)=O

Specifications

Melting Point 135.0°C to 138.0°C
Color Clear Colorless to Yellow
Assay Percent Range 97.5 % min. (HPLC)
Quantity 1 g
Solubility Information Solubility aq. soln.: 37 g/L (20°C).
Formula Weight 442.53
Chemical Name or Material Acepromazine maleate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.