missing translation for 'onlineSavingsMsg'
Learn More

Acephate, TRC

Product Code. 30134901
Change view
Click to view available options
Quantity:
50 mg
500 mg
1 g
Unit Size:
1g
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134901 500 mg 500mg
17957829 1 g 1g
17989179 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134901 Supplier Toronto Research Chemicals Supplier No. TRCA130850500MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30560-19-1
Molecular Formula C4 H10 N O3 P S
Molecular Weight (g/mol) 183.17
Synonym Phosphoramidothioic acid, acetyl-, O,S-dimethyl ester (8CI,9CI), Acefate, Acephate, Acetamidophos, Aciphid, Ant Hayter, Asataf, DMAP, Fire Ant Hayter Bait, Lepitect, O,S-Dimethyl N-acetylphosphoramidothioate, O,S-Dimethyl N-acetylphosphoramidothiolate, O,S-Dimethyl acetylphosphoramidothioate, Orthene, Orthene 75SP, Orthene 97, Orthene PCO, Orthene S, Orthene TTO, Orthene TTO 97, Orthene-755, Ortran, Payload, Precise, RE 12420
IUPAC Name N-[methoxy(methylsulfanyl)phosphoryl]acetamide
SMILES COP(=O)(NC(=O)C)SC

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C4H10NO3PS/c1-4(6)5-9(7,8-2)10-3/h1-3H3,(H,5,6,7)
Formula Weight 183.0119
Chemical Name or Material Acephate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.