missing translation for 'onlineSavingsMsg'
Learn More

Acenaphthenequinone, 95%

Product Code. 10677432 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10677432 25 g Glass bottle 25g
10579443 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10677432 Supplier Thermo Scientific Acros Supplier No. 102190250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 82-86-0 | C12H6O2 | 182.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82-86-0
Molecular Formula C12H6O2
Molecular Weight (g/mol) 182.18
MDL Number MFCD00003805
InChI Key AFPRJLBZLPBTPZ-UHFFFAOYSA-N
Synonym acenaphthenequinone, acenaphthoquinone, 1,2-acenaphthylenedione, acenaphthenedione, 1,2-acenaphthenequinone, 1,2-acenaphthenedione, acenaphthaquinone, acenaphthylene-1,2-quinone, acenaphthylenequinone, acenaphthodione
PubChem CID 6724
ChEBI CHEBI:15342
IUPAC Name acenaphthylene-1,2-dione
SMILES C1=CC2=C3C(=C1)C(=O)C(=O)C3=CC=C2

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 257.0°C to 263.0°C
Color Ochre
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Beilstein 07, 744
Solubility Information Solubility in water: insoluble. Other solubilities: slightly soluble in ethanol,benzene,soluble in ligroin
Formula Weight 182.18
Percent Purity 95%
Physical Form Powder or Granules
Chemical Name or Material Acenaphthenequinone
Show More Show Less
compliance-icons
Product Identifier
  • Acenaphthenequinone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.