missing translation for 'onlineSavingsMsg'
Learn More

Acenaphthene, 99%

Product Code. 10175113 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10175113 500 g Plastic bottle 500g
10201393 5 g Glass Bottle 5g
10741861 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10175113 Supplier Thermo Scientific Acros Supplier No. 201345000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 83-32-9 | C12H10 | 154.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83-32-9
Molecular Formula C12H10
Molecular Weight (g/mol) 154.21
MDL Number MFCD00003807
InChI Key CWRYPZZKDGJXCA-UHFFFAOYSA-N
Synonym acenaphthene, 1,8-ethylenenaphthalene, naphthyleneethylene, peri-ethylenenaphthalene, acenaphthylene, 1,2-dihydro, ethylenenaphthalene, 1,8-dihydroacenaphthalene, unii-v8ut1gac5y, ccris 5951, v8ut1gac5y
PubChem CID 6734
ChEBI CHEBI:22154
IUPAC Name 1,2-dihydroacenaphthylene
SMILES C1CC2=C3C1=CC=CC3=CC=C2

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 90°C to 95°C
Color Brown
Density 1.0600g/mL
Boiling Point 279°C
Flash Point 135°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Quantity 500 g
Beilstein 05, IV, 1834
Merck Index 15, 29
Specific Gravity 1.06
Solubility Information Solubility in water: insoluble. Other solubilities: 3.2g/100mL glacial acetic acid,1g/31mL alcohol- 1g/56mL methanol,1g/25mL propanol- 1g/2.5mL chloroform,1g/5mL benzene- 1g/5mL toluene
Formula Weight 154.21
Percent Purity 99%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material Acenaphthene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.