missing translation for 'onlineSavingsMsg'
Learn More

Acenaphthene (1,2-Dihydro Acenaphthylene), TRC

Product Code. 30385452
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30385452 10 g
30370466 100 g
30384895 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30385452 Supplier Toronto Research Chemicals Supplier No. TRCD44833010G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83-32-9
Molecular Formula C12 H10
Molecular Weight (g/mol) 154.21
Synonym Acenaphthene, 1,2-Dihydroacenaphthylene, peri-Ethylenenaphthalene, 1,8-Ethylenenaphthalene
IUPAC Name 1,2-dihydroacenaphthylene
SMILES C1Cc2cccc3cccc1c23

Specifications

Quantity 10 g
InChI Formula InChI=1S/C12H10/c1-3-9-4-2-6-11-8-7-10(5-1)12(9)11/h1-6H,7-8H2
Formula Weight 154.0783
Chemical Name or Material Acenaphthene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.