missing translation for 'onlineSavingsMsg'
Learn More

Aceglutamide, MedChemExpress

Product Code. 16452309
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16452309 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16452309 Supplier Medchem Express Supplier No. HYB1065
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Aceglutamide (α-N-Acetyl-L-glutamine) is a psychostimulant and nootropic, used to improve memory and concentration.

Aceglutamide (α-N-Acetyl-L-glutamine) is a psychostimulant and nootropic, used to improve memory and concentration.

Specifications

Chemical Name or Material Aceglutamide
CAS 2490-97-3
Color White
Purity Grade Notes Research
Molecular Formula C7H12N2O4
Synonym α-N-Acetyl-L-glutamine N2-Acetylglutamine
Solubility Information H2O : 33.33 mg/mL (177.12 mM; Need ultrasonic) ∣DMSO : 16.67 mg/mL (88.59 mM; Need ultrasonic)
SMILES O=C(N)CC[C@@H](C(O)=O)NC(C)=O
Molecular Weight (g/mol) 188.18
Quantity 1 g
Formula Weight 188.18
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.