missing translation for 'onlineSavingsMsg'
Learn More

Acefylline, MedChemExpress

Product Code. 16488776
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488776 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16488776 Supplier Medchem Express Supplier No. HYB1505
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Acefylline (Theophyllineacetic acid), a xanthine derivative, is an adenosine receptor antagonist. Acefylline is a peptidylarginine deiminase (PAD) activator. Acefylline is also a bronchodilator, which inhibits rat lung cAMP phosphodiesterase isoenzymes.

Acefylline (Theophyllineacetic acid), a xanthine derivative, is an adenosine receptor antagonist. Acefylline is a peptidylarginine deiminase (PAD) activator. Acefylline is also a bronchodilator, which inhibits rat lung cAMP phosphodiesterase isoenzymes.

Specifications

Chemical Name or Material Acefylline
CAS 652-37-9
Color White
Purity Grade Notes Research
Molecular Formula C9H10N4O4
Quantity 100 mg
Synonym Theophyllineacetic acid Theophylline-7-acetic acid
Solubility Information DMSO : 33.33 mg/mL (139.92 mM; Need ultrasonic)
SMILES O=C(O)CN1C=NC(N(C)C(N2C)=O)=C1C2=O
Molecular Weight (g/mol) 238.2
Formula Weight 238.2
Percent Purity 99.89%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.