missing translation for 'onlineSavingsMsg'
Learn More

Acedoben, TRC

Product Code. 30009120
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009120 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30009120 Supplier Toronto Research Chemicals Supplier No. TRCA1305005G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 556-08-1
Molecular Formula C9 H9 N O3
Synonym Benzoic acid, 4-(acetylamino)-, Benzoic acid, p-acetamido- (6CI,7CI,8CI), 4-(Acetylamino)benzoic acid, 4-Acetamidobenzoic acid, 4'-Carboxyacetanilide, Acedoben, N-Acetyl-p-aminobenzoic acid, NSC 4002, p-(Acetoamino)benzoic acid, p-(Acetylamino)benzoic aci
InChI Formula InChI=1S/C9H9NO3/c1-6(11)10-8-4-2-7(3-5-8)9(12)13/h2-5H,1H3,(H,10,11)(H,12,13)
SMILES CC(=O)Nc1ccc(cc1)C(=O)O
IUPAC Name 4-acetamidobenzoic acid
Molecular Weight (g/mol) 179.173
Formula Weight 179.058
Percent Purity >95%
Quantity 5 g
Chemical Name or Material Acedoben
Grade HPLC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.