missing translation for 'onlineSavingsMsg'
Learn More

Aceclofenac, 99%

Product Code. 15463139
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15463139 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15463139 Supplier Thermo Scientific Alfa Aesar Supplier No. J60355.06
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 89796-99-6 | C16H13Cl2NO4 | 354.183 g/mol

A non-steroidal, anti-inflammatory drug (NSAID) with potent inhibitory activity in several models of inflammation

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

A non-steroidal, anti-inflammatory drug (NSAID) with potent inhibitory activity in several models of inflammation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89796-99-6
Molecular Formula C16H13Cl2NO4
Molecular Weight (g/mol) 354.183
MDL Number MFCD00864296
InChI Key MNIPYSSQXLZQLJ-UHFFFAOYSA-N
Synonym aceclofenac, aceclofenaco, aceclofenacum, preservex, airtal, falcol, gerbin, aceclofar, aceclofenacum latin, aceclofenaco spanish
PubChem CID 71771
ChEBI CHEBI:31159
IUPAC Name 2-[2-[2-(2,6-dichloroanilino)phenyl]acetyl]oxyacetic acid
SMILES C1=CC=C(C(=C1)CC(=O)OCC(=O)O)NC2=C(C=CC=C2Cl)Cl

Specifications

Melting Point 149°C to 153°C
Color White
UN Number UN2811
Merck Index 14,22
Solubility Information Soluble in DMSO
Quantity 5 g
Formula Weight 354.18
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Aceclofenac

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.