missing translation for 'onlineSavingsMsg'
Learn More

Acebutolol hydrochloride, Dr. Ehrenstorfer

Code produit 30313827
Modifier l'affichage
Click to view available options
Quantity:
50 mg
Conditionnement:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantity Conditionnement
30313827 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 30313827 Fournisseur Dr. Ehrenstorfer Code fournisseur DREC10005150
 Restricted Item
Estimated Shipment: 30-11-2026
Ajouter au panier
Ajouter au panier

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Spécification

Chemical Name or Material Acebutolol Hydrochloride
CAS 34381-68-5
Analyte or Component Names Acebutolol Hydrochloride
Shipping Condition +20°C
Molecular Formula C18 H28 N2 O4 . Cl H
Certifications/Compliance ISO/IEC 17025
Synonym Acebutolol Hydrochloride
InChI Formula InChI=1S/C18H28N2O4.ClH/c1-5-6-18(23)20-14-7-8-17(16(9-14)13(4)21)24-11-15(22)10-19-12(2)3;/h7-9,12,15,19,22H,5-6,10-11H2,1-4H3,(H,20,23);1H
SMILES Cl.CCCC(=O)Nc1ccc(OCC(O)CNC(C)C)c(c1)C(=O)C
IUPAC Name N-[3-acetyl-4-[2-hydroxy-3-(propan-2-ylamino)propoxy]phenyl]butanamide;hydrochloride
Molecular Weight (g/mol) 372.89
Quantity 50 mg
Formula Weight 372.1816 g/mol
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.