missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ACE Recombinant Rabbit Monoclonal Antibody (K01_1M75)

Product Code. 30829925
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30829925 100 μL
30827476 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30829925 Supplier Invitrogen™ Supplier No. MA563309
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: PLPDNYPEGI DLETDEAKAD RFVEEYDRTA QVLLNEYAEA NWQYNTNITI EGSKILLEKS TEVSNHTLKY GTRAKTFDVS NFQNSSIKRI IKKLQNLDRA VLPPKELEEY NQILLDMETT YSLSNICYTN GTCMPLEPDL TNMMATSRKY EELLWAWKSW RDKVGRAILP FFPKYVEFSN KIAKLNGYTD AGDSWRSLYE SDNLEQDLEK LYQELQPLYL NLHAYVRRSL HRHYGSEYIN LDGPIPAHLL GNMWAQTWSN IYDLVAPFPS APNIDATEAM IKQGWTPRRI FKEADNFFTS LGLLPVPPEF WNKSMLEKPT DGREVVCHPS AWDFYNGKDF RIKQCTSVNM EDLVIAHHEM GHIQYFMQYK DLPVTFREGA NPGFHEAIGD IMALSVSTPK HLYSLNLLST EGSGYEYDIN FLMKMALDKI AFIPFSYLID QWRWRVFDGS ITKENYNQEW WSLRLKYQGL CPPVPRSQGD FDPGSKFHVP ANVPYVRYFV SFIIQFQFHE ALCRAAGHTG PLHKCDIYQS KEAGKLLADA MKLGYSKPWP EAMKLITGQP NMSASAMMNY FKPLTEWLVT ENRRHGETLG WPEYNWAPNT AR Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 94%, Pig - 81%, Cynomolgus monkey - 82%.

This gene encodes an enzyme involved in catalyzing the conversion of angiotensin I into a physiologically active peptide angiotensin II. Angiotensin II is a potent vasopressor and aldosterone-stimulating peptide that controls blood pressure and fluid-electrolyte balance. This enzyme plays a key role in the renin-angiotensin system. Many studies have associated the presence or absence of a 287 bp Alu repeat element in this gene with the levels of circulating enzyme or cardiovascular pathophysiologies. Two most abundant alternatively spliced variants of this gene encode two isozymes-the somatic form and the testicular form that are equally active. Multiple additional alternatively spliced variants have been identified but their full length nature has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACE
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1M75
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 150mM NaCl, 0.05% BSA, 40% glycerol and 0.01% sodium azide; pH 7.4
Gene Ace
Gene Accession No. P09470, P47820
Gene Alias ACE; ACE1; ACE-T; angiotensin 1 converting enzyme 1; angiotensin converting enzyme; angiotensin converting enzyme, somatic isoform; angiotensin I converting enzyme; angiotensin I converting enzyme (peptidyl-dipeptidase A) 1; angiotensin I converting enzyme 1; angiotensin I converting enzyme peptidyl-dipeptidase A 1 transcript; angiotensin I-converting enzyme (Dipeptidyl carboxypeptidase 1); angiotensin-converting enzyme; Angiotensin-converting enzyme, soluble form; angiotensin-converting enzyme; LOW QUALITY PROTEIN: angiotensin-converting enzyme; angiotensin-converting enzyme-like; AW208573; carboxycathepsin; CD143; CD143 antigen; DCP; Dcp1; dipeptidyl carboxypeptidase 1; Dipeptidyl carboxypeptidase 1 (Angiotensin I-converting enzyme); dipeptidyl carboxypeptidase I; dipeptidyl peptidase; I79_009670; ICH; kininase II; LOC103158533; MGC26566; MVCD3; peptidase P; peptidyl dipeptidase i; StsRR92; testicular ECA
Gene Symbols Ace
Host Species Rabbit
Immunogen Recombinant protein of mouse Angiotensin Converting Enzyme 1 (aa 636-1237).
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11421, 24310
Target Species Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.