missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACE Monoclonal Antibody (536CT24.1.3)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA537594
This item is not returnable.
View return policy
Description
ACE Monoclonal Antibody for Western Blot
This gene encodes an enzyme involved in catalyzing the conversion of angiotensin I into a physiologically active peptide angiotensin II. Angiotensin II is a potent vasopressor and aldosterone-stimulating peptide that controls blood pressure and fluid-electrolyte balance. This enzyme plays a key role in the renin-angiotensin system. Many studies have associated the presence or absence of a 287 bp Alu repeat element in this gene with the levels of circulating enzyme or cardiovascular pathophysiologies. Two most abundant alternatively spliced variants of this gene encode two isozymes-the somatic form and the testicular form that are equally active. Multiple additional alternatively spliced variants have been identified but their full length nature has not been determined.
Specifications
| ACE | |
| Monoclonal | |
| Conc. Not Determined | |
| ascites with 0.09% sodium azide; pH 7.4 | |
| P09470, P12821 | |
| Ace | |
| KLH conjugated synthetic peptide between 1274-1306 amino acids from human ACE. | |
| RUO | |
| 11421, 1636 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 536CT24.1.3 | |
| Unconjugated | |
| Ace | |
| ACE; ACE1; ACE-T; angiotensin 1 converting enzyme 1; angiotensin converting enzyme; angiotensin converting enzyme, somatic isoform; angiotensin I converting enzyme; angiotensin I converting enzyme (peptidyl-dipeptidase A) 1; angiotensin I converting enzyme 1; angiotensin I converting enzyme peptidyl-dipeptidase A 1 transcript; angiotensin I-converting enzyme (Dipeptidyl carboxypeptidase 1); angiotensin-converting enzyme; Angiotensin-converting enzyme, soluble form; angiotensin-converting enzyme; LOW QUALITY PROTEIN: angiotensin-converting enzyme; angiotensin-converting enzyme-like; AW208573; carboxycathepsin; CD143; CD143 antigen; DCP; Dcp1; dipeptidyl carboxypeptidase 1; Dipeptidyl carboxypeptidase 1 (Angiotensin I-converting enzyme); dipeptidyl carboxypeptidase I; dipeptidyl peptidase; I79_009670; ICH; kininase II; LOC103158533; MGC26566; MVCD3; peptidase P; peptidyl dipeptidase i; StsRR92; testicular ECA | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction