missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ACE-2 Monoclonal specifically detects ACE-2 in Human samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | ACE-2 |
| Applications | Flow Cytometry, ELISA, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | AC384 |
| Conjugate | FITC |
| Dilution | Flow Cytometry, ELISA |
| Formulation | PBS |
| Gene Alias | ACEHangiotensin I converting enzyme 2, ACE-related carboxypeptidase, angiotensin I converting enzyme (peptidyl-dipeptidase A) 2, angiotensin-converting enzyme 2, Angiotensin-converting enzyme homolog, DKFZp434A014, EC 3.4.17, EC 3.4.17.23, Metalloprotease MPROT15 |
| Host Species | Mouse |
| Immunogen | Recombinant human ACE-2. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?