missing translation for 'onlineSavingsMsg'
Learn More

ACE-2 Antibody (AC18F), Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18371990 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18371990 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18371990 Supplier Novus Biologicals Supplier No. NBP280035AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ACE-2 Monoclonal specifically detects ACE-2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACE-2
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone AC18F
Conjugate Alexa Fluor 350
Dilution Western Blot, Flow Cytometry, ELISA
Formulation 50mM Sodium Borate
Gene Alias ACEHangiotensin I converting enzyme 2, ACE-related carboxypeptidase, angiotensin I converting enzyme (peptidyl-dipeptidase A) 2, angiotensin-converting enzyme 2, Angiotensin-converting enzyme homolog, DKFZp434A014, EC 3.4.17, EC 3.4.17.23, Metalloprotease MPROT15
Host Species Mouse
Immunogen Recombinant human ACE-2.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 59272
Test Specificity Recognizes human ACE-2. Does not detect recombinant human ACE-2 that has a tag at the C-terminus of the protein.
Target Species Human, Hamster
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.