missing translation for 'onlineSavingsMsg'
Learn More

ACD Antibody (OTI1A2), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-01851

Product Code. 18062267

  • £439.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ACD Monoclonal specifically detects ACD in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ACD
Monoclonal
1 mg/mL
Western Blot 1:2000, Flow Cytometry 1:100, Immunocytochemistry/Immunofluorescence 1:100
ACD
Full length human recombinant protein of human ACD(NP_001075955) produced in HEK293T cell.
Affinity Purified
RUO
65057
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
OTI1A2
Unconjugated
adrenocortical dysplasia homolog (mouse), Pip1, PIP1Tint1, POT1 and TIN2 organizing protein, POT1 and TIN2-interacting protein, Ptop, PTOPTpp1, TIN2 interacting protein 1, TINT1adrenocortical dysplasia protein homolog, TPP1
Mouse
57.6 kDa
0.1 mL
Primary
Human
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only