Learn More
STEMCELL Technologies ACCUTASE™
Description
- A cell detachment solution of proteolytic and collagenolytic enzymes
- Useful for the routine detachment of cells from standard tissue culture plasticware and adhesion coated plasticware
- ACCUTASE™ does not contain mammalian or bacterial-derived products
- Each lot of ACCUTASE™ is tested for sterility (by USP membrane filtration method), enzymatic activity (tested with synthetic chromagenic tetrapeptides) and cell detachment from tissue culture plastic
Specifications
Specifications
| Cell Type | Neural Cells, PSC-Derived, Pluripotent Stem Cells |
| Quantity | 100 mL |