Learn More
STEMCELL Technologies ACCUTASE™
Product Code. 17189911
Encompass_Preferred
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Product Code. 17189911
Brand: STEMCELL Technologies 07922
Cell detachment solution
  • A cell detachment solution of proteolytic and collagenolytic enzymes
  • Useful for the routine detachment of cells from standard tissue culture plasticware and adhesion coated plasticware
  • ACCUTASE™ does not contain mammalian or bacterial-derived products
  • Each lot of ACCUTASE™ is tested for sterility (by USP membrane filtration method), enzymatic activity (tested with synthetic chromagenic tetrapeptides) and cell detachment from tissue culture plastic
Specifications
Cell Type Neural Cells, PSC-Derived, Pluripotent Stem Cells
Quantity 500 mL