missing translation for 'onlineSavingsMsg'
Learn More

AccuSEQ™ Real-Time PCR Detection Software, version 2.1.2

Product Code. 18713644 Shop All Applied Biosystems Products
Click to view available options
Quantity:
1 each
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 18713644

Brand: ABI Applied Biosystems A47130

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Content And Storage MycoSEQ, Probe, resDNASEQ
For Use With (Equipment) 7500 Fast System
Product Line AccuSEQ
Software Type Real-Time PCR Software
Product Type Real Time PCR Detection Software
For Use With (Application) Real Time PCR (qPCR)
Shipping Condition Room Temperature
Quantity 1 each
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.