Learn More
Applied Biosystems™ AccuSEQ™ Real-Time PCR Detection Software v4.0
Product Code. 31034143 Shop All Applied Biosystems Products
Change view
Click to view available options
Software Type:
Real-Time PCR Software
Real-Time PCR Software (Seat License)
Real-Time PCR Software (Upgrade License)
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Software Type
31034143 Real-Time PCR Software
31034550 Real-Time PCR Software (Seat License)
31034426 Real-Time PCR Software (Upgrade License)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 31034143 Supplier Applied Biosystems™ Supplier No. A40005162
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
AccuSEQ Real-Time PCR Detection Software is part of an integrated workflow for impurity and contaminant testing during biopharmaceutical manufacturing using the SEQ family of real-time PCR assays and helps support customers in meeting their local regulations, including 21 CFR Part 11 compliance.
AccuSEQ Real-Time PCR Detection Software v4.0 is part of an integrated workflow for impurity and contaminant testing during biopharmaceutical manufacturing using the SEQ family of real-time PCR assays, including MycoSEQ, SteriSEQ, resDNASEQ, and ViralSEQ assays. The software is designed to work with the QuantStudio 5 and QuantStudio 7 Pro real-time PCR systems with 96-well plates (0.1-mL, 0.2-mL) and 384-well plates for either fast or standard cycling. This software is provided via a link for online download.
Features of AccuSEQ Real-Time PCR Detection Software include:
  • Accurate host cell impurities quantitation for resDNASEQ host cell residual DNA quantitation assays
  • Automated presence/absence calls for MycoSEQ Mycoplasma, MycoSEQ Plus Mycoplasma, ViralSEQ MMV, and ViralSEQ Vesivirus assays
  • Accurate quantitation of Sf-rhabdovirus genomic RNA for ViralSEQ Quantitative Sf-Rhabdovirus Kit
  • Single software platform for all current and future SEQ real-time PCR assays
  • Security, audit, and e-signature capabilities to help enable 21 CFR Pt 11 compliance
  • Integrated, easy-to-use interface allows for consistency across multiple data applications and reduces the time needed to train your team
  • Flexible throughput of 10–500 samples per week
  • Complete traceability with assay-specific output, eliminating manual calculations
  • Comprehensive data management and reporting features
Residual DNA Quantitation
For host cell residual DNA quantitation, a custom module with next-generation algorithms can be used with the resDNASEQ residual host cell DNA quantitation assay for accurate quantitation.
Mycoplasma Detection
For rapid and accurate Mycoplasma detection, AccuSEQ software offers a plug-in analysis module for automated presence/absence calls when using the MycoSEQ Mycoplasma detection system. Automated analysis tools enable one-click processing of MycoSEQ assay data, helping deliver presence or absence calls within seconds of data collection being completed.
Rapid Sterility Testing
For rapid sterility testing, AccuSEQ software offers a plug-in analysis module for automated presence/absence calls when using the SteriSEQ Rapid Sterility Testing System. Automated analysis tools enable one-click processing of SteriSEQ™ Rapid Sterility Testing Kit (Cat. No. A57186) data, helping deliver presence or absence calls within seconds of data collection being completed.
MMV and Vesivirus Detection
AccuSEQ software is optimized and validated for the ViralSEQ MMV and ViralSEQ Vesivirus assays. The software's visual result summary provides immediate understanding of presence or absence of virus in your samples.
Sf-rhabdovirus Genomic RNA Quantitation
AccuSEQ software helps enable rapid and easy analysis of ViralSEQ Quantitative Sf-Rhabdovirus kit data without the hassle of multiple exporting steps and reliance on third party software.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) QuantStudio™ qPCR Systems
Product Line AccuSEQ
Software Type Real-Time PCR Software
Product Type Real Time PCR Detection Software
For Use With (Application) Real Time PCR (qPCR)