missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ accumet™ Glass Body Standard Size Combination Electrodes - Mercury-Free

For routine pH and titration measurements with small samples and small vessels, where separate half-cell electrodes may be impractical

Brand:  Fisherbrand™

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 11530064

  • £221.00 / Each
Estimated Shipment: 04-07-2024
to see stock.
Add to basket

Includes: Removable bulb protector

Description

Description

For routine pH and titration measurements with small samples and small vessels, where separate half-cell electrodes may be impractical

 

  • 0 to 14pH range
  • Porous ceramic junction
  • Twist-open/twist-close cap for easy filling
  • Constant distance between FS-5 Universal Glass pH bulb and reference junction means high reproducibility
  • Immersion depth, ∽4 in. (10.1cm)
  • 30 in. long (76.2cm) leads, except 13-620-91, which has 60 in. long (152.4cm) lead

 

Choice of Ag/AgCl Single- or Double-Junction internal reference elements

  • Single-junction models with Ag/AgCl reference are perfect for routine pH and titration measurements of aqueous solutions with small samples, small vessels
  • Double-junction models with Ag/AgCl reference are useful in aqueous and nonaqueous solutions, and in applications requiring TRIS buffers
  • For aqueous and nonaqueous samples
  • Double-junction Ag/AgCl eliminate measurement and clogging problems when used in research with Tris buffers

 

Specifications

Specifications

Twist Open/Twist Close
U.S. Standard/Pin
Glass
23°C to 212°C
SP135-500
pH meter
Ag/AgCl
-5°F to +100°F
1 Year
60 in.
152.4 cm
Blue
Single Ceramic
0 to 14
pH Combination Electrode
For routine pH and titration measurements with small samples and small vessels, where separate half cell electrodes may be impractical
Removable bulb protector
<10 sec.
-5°C to +100°C
4 in.
101.6 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers