Learn More
Invitrogen™ Accumax - Cell Aggregate Dissociation Medium
Description
Accumax has been shown effective in dissociating cell clumps in a variety of cell lines including hybridomas, CHO, BHK, 293, COS and Sf9 cells. The use of Accumax may effect the integrity of certain antigenic epitopes resulting in diminished antibody staining. This should be determined experimentally for individual antibody clones. Thaw Accumax at room temperature or 4°C.
Recommended Storage
Store at −20°C. Thawing during shipment will not affect performance.
Especificaciones
Especificaciones
| Cell Type | Mammalian Cells |
| Form | Liquid |
| Product Type | Cell Culture Dissociation Reagent |
| Without Additives | No Phenol Red |
| Quantity | 100 mL |
For Research Use Only. Not for use in diagnostic procedures.
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.