Learn More
STEMCELL Technologies ACCUMAX™
Cell detachment solution
Brand:  STEMCELL Technologies 07921
Additional Details : Weight : 0.10000kg
Product Code. 17179911
  • £207.90 / 100mL
Estimated Shipment: 07-05-2024
to see stock.
Description
Description
  • ACCUMAX™ is a cell detachment solution of proteolytic and collagenolytic enzymes
  • It is useful for creating single-cell suspensions from clumped cell cultures for accurate cell counting, detachment of cells from primary tissue, and the routine detachment of cells from standard tissue culture plasticware and adhesion-coated plasticware
  • ACCUMAX™ does not contain mammalian- or bacterial-derived products
  • Each lot of ACCUMAX™ is tested for sterility (by USP membrane filtration method), enzymatic activity (tested with synthetic chromagenic tetrapeptides), and cell detachment from tissue culture plastic
Specifications
Specifications
Pluripotent Stem Cells
100 mL