missing translation for 'onlineSavingsMsg'
Learn More

BD Biosciences™ Accudrop Beads (RUO)

Product Code. 15889881
Click to view available options
Diameter (Metric):
7.5 μm
Unit Size:
25 tests
This item is not returnable. View return policy

Product Code. 15889881

Brand: BD Biosciences™ 661612

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Suitable for setting the drop delay on BD cell sorters.

  • Set the drop delay on BD cell sorters.
  • Contains a single population of 7.5-μm particles.
  • Every particle contains a fluorophore that is excited at 660 nm and emits at 780 nm.
  • Particles are supplied in 1.5 mL of water containing 0.05% Tween™ 20 and 2 mM sodium azide.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1.0 x 108 beads/mL
Content And Storage Store vials at 2°C–8°C. Protect from exposure to light.
Contents Particles supplied in Water, 0.05% Tween 20, 2 mM sodium azide
No. of Tests 25
Quantity 1.5 mL
Stability Each reagent is stable until the expiration date shown on the bottle label when stored as directed. Use diluted particles within 24 hours after dilution.
Uniformity Single population size
Diameter (Metric) 7.5 μm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.