Learn More
Thermo Scientific™ Accucore™ 150-C4 HPLC Columns
Use this specially designed solid core column with a C4 functionality for biomolecules that bind too tightly to C8 or C18 columns.
£388.62 - £962.88
Specifications
For Use With | HPLC |
---|---|
Particle Size | 2.6 μm |
pH | 2 to 9 |
Pore Size | 150 Å |
Surface Area | 80 m²/g |
Product Code | Brand | Particle Size | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Particle Size | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||
12685695
|
Thermo Scientific™
16526-032130 |
2.6 μm | 2.1 mm | 30 mm |
£506.89
Each |
In Stock
Log in to see stock. |
|||
12615705
|
Thermo Scientific™
16526-052130 |
2.6 μm | 2.1 mm | 50 mm |
£617.95
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||
12645705
|
Thermo Scientific™
16526-102130 |
2.6 μm | 2.1 mm | 100 mm |
£725.22
Each |
Estimated Shipment: 25-06-2024
Log in to see stock. |
|||
12655705
|
Thermo Scientific™
16526-103030 |
2.6 μm | 3 mm | 100 mm |
£725.22
Each |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||
12665705
|
Thermo Scientific™
16526-104630 |
2.6 μm | 4.6 mm | 100 mm |
£725.22
Each |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||
12685705
|
Thermo Scientific™
16526-152130 |
2.6 μm | 2.1 mm | 150 mm |
£835.32
Each |
In Stock
Log in to see stock. |
|||
12695705
|
Thermo Scientific™
16526-154630 |
2.6 μm | 4.6 mm | 150 mm |
£835.32
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||
12655715
|
Thermo Scientific™
16526-157569 |
2.6 μm | 0.075 mm | 150 mm |
£946.82
Each |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||
12605715
|
Thermo Scientific™
16526-012105 |
2.6 μm | 2.1 mm | 10 mm |
£470.86
Pack of 4 |
In Stock
Log in to see stock. |
|||
12615715
|
Thermo Scientific™
16526-013005 |
2.6 μm | 3 mm | 10 mm |
£470.86
Pack of 4 |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||
12625715
|
Thermo Scientific™
16526-014005 |
2.6 μm | 4 mm | 10 mm |
£470.86
Pack of 4 |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||
12665715
|
Thermo Scientific™
16526-507569 |
2.6 μm | 0.075 mm | 500 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12675705
|
Thermo Scientific™
16526-153030 |
2.6 μm | 3 mm | 150 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12695695
|
Thermo Scientific™
16526-033030 |
2.6 μm | 3 mm | 30 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12625705
|
Thermo Scientific™
16526-053030 |
2.6 μm | 3 mm | 50 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Offer lower hydrophobic retention, ideal for optimal retention of proteins and larger peptides with Thermo Scientific™ Accucore™ 150-C4 LC Columns. Based on Core Enhanced Technology, Thermo Scientific Accucore columns provide fast, high-resolution biomolecule separations without the elevated backpressures required by sub-2μm particles.
- 150Å pore size solid core particles for fast biomoleucule separations
- Superb resolution at low backpressures
- Exceptionally rugged columns
- Significantly lower hydrophobic retention than C18
- Ideal for optimal retention of proteins and larger peptides
Ordering Information:
Other custom column dimensions are available. Please call your local Customer Service for more information.
Specifications
HPLC | |
2 to 9 | |
80 m²/g | |
L26 | |
Spherical, Solid Core, Ultrapure Silica | |
Reversed Phase | |
Reversed Phase |
2.6 μm | |
150 Å | |
60°C | |
C4 | |
2% | |
Yes |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.