missing translation for 'onlineSavingsMsg'
Learn More

ACCN2 Antibody (S271-44), Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18423666 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18423666 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18423666 Supplier Novus Biologicals Supplier No. NBP222409JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ACCN2 Monoclonal antibody specifically detects ACCN2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACCN2
Applications Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone S271-44
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias Acid-sensing ion channel 1, acid-sensing ion channel 1a protein, amiloride-sensitive cation channel 2, neuronal, ASIC, ASIC1ASIC1A, BNAC2, BNaC2ACCN2 variant 3, Brain sodium channel 2, Cation channel, amiloride-sensitive, neuronal, 2, hBNaC2
Host Species Mouse
Immunogen Fusion protein amino acids 460-526 (Cytoplasmic C-terminus) of mouse ASIC1
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 41
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.