missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACCN2 Antibody (S271-44), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22409AF488
This item is not returnable.
View return policy
Description
ACCN2 Monoclonal specifically detects ACCN2 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| ACCN2 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ASIC1 | |
| Fusion protein amino acids 460-526 (Cytoplasmic C-terminus) of mouse ASIC1 | |
| 0.1 mL | |
| 41 | |
| Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| S271-44 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Acid-sensing ion channel 1, acid-sensing ion channel 1a protein, amiloride-sensitive cation channel 2, neuronal, ASIC, ASIC1ASIC1A, BNAC2, BNaC2ACCN2 variant 3, Brain sodium channel 2, Cation channel, amiloride-sensitive, neuronal, 2, hBNaC2 | |
| Mouse | |
| Protein G purified | |
| Primary | |
| Detects approx 60kDa. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction