missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACCN1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92023-0.1ml
This item is not returnable.
View return policy
Description
ACCN1 Polyclonal antibody specifically detects ACCN1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| ACCN1 | |
| Polyclonal | |
| Western Blot 1:1000-1:2000 | |
| Acid-sensing ion channel 2, Amiloride-sensitive brain sodium channel, amiloride-sensitive cation channel 1, neuronal, ASIC2a, ASIC2Mammalian degenerin homolog, BNAC1, BNaC1ACCN, BNC1Amiloride-sensitive cation channel neuronal 1, degenerin, hBNaC1, MDEGBrain sodium channel 1, neuronal amiloride-sensitive cation channel 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 120-400 of human ASIC2 (NP_001085.2). NLQIPDPHLADPSVLEALRQKANFKHYKPKQFSMLEFLHRVGHDLKDMMLYCKFKGQECGHQDFTTVFTKYGKCYMFNSGEDGKPLLTTVKGGTGNGLEIMLDIQQDEYLPIWGETEETTFEAGVKVQIHSQSEPPFIQELGFGVAPGFQTFVATQEQRLTYLPPPWGECRSSEMGLDFFPVYSITACRIDCETRYIVENCNCRMVHMPGDAPFCTPEQHKECAEPALGLLAEKDSNYCLCRTPCNLTRYNKELSMVKIPSKTSAKYLEKKFNKSEKYISE | |
| 0.1 mL | |
| Apoptosis, Cell Biology, Neuroscience | |
| 40 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction