missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Acclaim™ Trinity P1 HPLC Columns

Product Code. 11362073 Shop All Thermo Scientific Products
Change view
Click to view available options
Length (Metric):
10 mm
50 mm
100 mm
150 mm
Diameter (Metric):
2.1 mm
3 mm
Unit Size:
Each
Pack of 2
Product Code. Length (Metric) Diameter (Metric) unitSize
11362073 150 mm 3 mm Each
11371943 100 mm 3 mm Each
11381943 50 mm 3 mm Each
11391943 100 mm 2.1 mm Each
11372073 150 mm 2.1 mm Each
11382073 50 mm 2.1 mm Each
11321953 10 mm 2.1 mm Pack of 2
11301953 10 mm 3 mm Each
8 options
This item is not returnable. View return policy

Product Code. 11362073

Brand: Thermo Scientific™ 075563

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Use these mixed-mode columns for simultaneous separation of pharmaceutical drug substances and their counter ions with high efficiency.

Simultaneously separate pharmaceutical drug substances and their counterions with high-efficiency Thermo Scientific™ Acclaim™ Trinity P1 LC Columns. A unique trimodal surface chemistry provides concurrent reversed-phase, cation-exchange and anion-exchange functionalities for maximum flexibility in method development. Easily optimize selectivity by adjusting mobile phase buffer concentration, pH and solvent content—either jointly or independently—to accelerate chromatographic separations and increase productivity.

The Acclaim Trinity P1 stationary phase, based on Nanopolymer Silica Hybrid (NSH) technology, consists of high-purity porous, spherical 3μm silica particles, coated with charged nanopolymer beads. The unique surface chemistry includes an inner-pore area modified with an organic layer that provides both reversed-phase and anion-exchange properties. The outer-pore surface, conversely, is modified with cation-exchange functionality for unparalleled chromatographic performance.

Superior Performance for Pharmaceutical Applications

  • Ideal selectivity for simultaneous separation of Active Pharmaceutical Ingredients (API) and counterions (basic, neutral and acidic analytes)
  • Effective retention of hydrophilic ionic and ionizable analytes without ion-pairing reagents
  • Distinctive spatial separation of the anion-exchange and cation-exchange regions
  • Flexible method development with each retention mechanism controlled independently
  • Low column bleed; compatible with mass spectrometry (MS), charged aerosol detection (CAD) and evaporative light scattering detectors (ELSD)
  • Available in 2.1 and 3.0mm diameters and 50, 100 and 150mm lengths

Complement the Acclaim Trinity P1 column with the Acclaim Trinity P2 column to provide a total solution for pharmaceutical counterion analysis by HPLC. Use the Acclaim Trinity P2 column to separate counterions, including monovalent and divalent cations or anions.

About the Family of Acclaim Specialty HPLC Columns
Acclaim specialty columns have been developed, manufactured and tested to meet the separation needs of specific applications. These columns are based on novel and unique chemistries and provide superior resolution with ease of use.

  • Use tested to guarantee consistent separations
  • Ideal selectivity for separating specific target analytes
  • Excellent peak shapes
  • Performance tested to ensure high quality and reliability for specific chromatographic separations
TRUSTED_SUSTAINABILITY

Specifications

Column Format Analytical Column
Diameter (Metric) 3 mm
For Use With ELSD, UV Detector
Length (Metric) 150 mm
Max. Pressure 4500 psi (310 bar)
Particle Size 3 μm
pH 2.5 to 7
Pore Size 300 Å
Quantity Each
Surface Area 100 m²/g
Temperature 45°C
Stationary Phase Reversed Phase/Anion Exchange/Cation Exchange
Product Line Acclaim
Packing Material Spherical, Nanopolymer Hybrid Silica
Column Type Mixed Mode
Endcapped Proprietary
Phase Reversed Phase and Ion Exchange
Type Analytical Column
Green Features Responsibly packaged
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.