Learn More
Thermo Scientific™ Acclaim™ SEC-1000 Size Exclusion Chromatography HPLC Columns
Shop All Thermo Scientific ProductsDescription
Separate water-soluble polymers and oligomers with the Acclaim family of resin-based, high-performance, size-exclusion chromatography columns.
Separate water-soluble polymers and oligomers with Thermo Scientific™ Acclaim™ SEC-1000 Size Exclusion HPLC Columns. These resin-based, high-performance, size-exclusion chromatography columns cover molecular weight range: 7μm, 1000Å. The combination of a proprietary mono-dispersed multi-pore hydrophilic resin and small particle size provides high-resolution and good linearity of the calibration curve with no inflection points for accurate, reliable analysis of a variety of water-soluble polymers.
High-Performance Analysis of Water-Soluble Polymers
- High-resolution analysis at reduced solvent consumption in available 4.6 x 300mm dimension
- Stable surface bonding leads to low column bleed and enhances compatibility with UV, RI, MS, ELSD and Thermo Scientific Dionex™ Corona™ Charged Aerosol Detectors
Specifications
Specifications
| Column Format | Guard Column |
| Diameter (Metric) | 4.6 mm |
| Length (Metric) | 33 mm |
| Particle Shape | Spherical |
| Particle Size | 7 μm |
| pH | 2 to 12 |
| Pore Size | 1000 Å |
| Quantity | Each |
| Surface Area | 1000 m²/g |
| Temperature | 60°C |
| Show More |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.