missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Acclaim™ PepMap™ C8 Peptide Analysis HPLC Column, 3 μm, 0.075 mm x 150 mm
Column Format: Nanoviper; Carbon Load: 9%; Pore Size: 100 Å; Column Phase: Reversed Phase
Brand: Thermo Scientific™ 164706
Code : A0
Additional Details : Weight : 0.20000kg
Description
- Ease of use, no sleeve for small capillaries
- Tool-free assembly
- No column damage from overtightening: capable of withstanding pressures up to 1000 bars and is compatible with third party valves and unions
- No experimental failure due to bad connections
Specifications
Nanoviper | |
150 mm | |
3 μm | |
L7 | |
nanoLC | |
9% | |
Reversed Phase |
0.075 mm | |
14500 psi (1000 bar) | |
100 Å | |
C8 | |
Silica | |
Peptide Analysis |