missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Support the enrichment of phosphopeptides with Thermo Scientific™ Titanium-Dioxide Nano-Trap Columns.
Specifications
Specifications
| Column Format | Trap Column |
| Diameter (Metric) | 0.2 mm |
| Length (Metric) | 10 mm |
| Max. Pressure | 7250 psi (500 bar) |
| Particle Size | 5 μm |
| Stationary Phase | Titanium Dioxide |
| Product Line | nanoLC |
| Packing Material | Titanium Dioxide |
| Column Type | Reversed Phase |
| Phase | Reversed Phase |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?