missing translation for 'onlineSavingsMsg'
Learn More

Miele Professional™ Accessory large washer-disinfector

Product Code. 11675980 Shop All Miele Products
Change view
Click to view available options
Color:
Stainless Steel
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
11675980 Stainless Steel Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11675980 Supplier Miele Professional™ Supplier No. 5746210
Add to basket
Add to basket
This item is not returnable. View return policy

Lid A 19 for fixing delicate wash items (PG 8527)

  • Perforated plate 7 x 7 x 3 mm
  • Suitable for insert E 969
TRUSTED_SUSTAINABILITY

Specifications

Material Stainless Steel
Product Type Cover
Color Stainless Steel
For Use With (Equipment) Washer-disinfectors, laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.