Learn More
Cytiva Accessory Kits
Product Code. 11633224 Shop All Cytiva Products
Change view
Click to view available options
For Use With:
For HiScale 26 Columns
For HiScale 50 Columns
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With unitSize
11633224 For HiScale 26 Columns Each
11643224 For HiScale 50 Columns Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11633224 Supplier Cytiva Supplier No. 28966374
Only null left
Add to basket
Add to basket
Specifications
Type Accessory Kits
Packaging Quantity 1 Each
Includes Net ring, support screen, o-ring, two ferrules and two finger tight stop plugs
For Use With For HiScale 26 Columns