Learn More
Miele Professional™ Accessory Booster Pump PLW8636 APLW 881
Product Code. 30533236 Shop All Miele Products
Click to view available options
Rack Type:
Rec out
Unit Size:
1 sheet
Product Code. 30533236
Brand: Miele Professional™ 12426550
Rec in/out water inlet and drain for PLW 8636
  • Rec in/out water inlet and drain for PLW 8636
  • Required in order to pump a medium out of a buffer container
  • For use with an existing on-site recycling tank
  • Can be used with existing on-site container, e.g. IBC tank
  • Installed by Customer Service
TRUSTED_SUSTAINABILITY
Specifications
Product Type Booster Pump
Rack Type Rec out
For Use With (Equipment) Washer-Disinfectors
Quantity 1 Ea.