missing translation for 'onlineSavingsMsg'
Learn More

Merck Accessories for YY3009000 Filter Holder, 90 mm

Accessories for filter holders for sample preparation.

Brand:  Merck YY2214257

79.65 GBP valid until 2024-12-31
Use promo code "23269" to get your promotional price.



Code : 72

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10672632

  • £85.50 / Each
Estimated Shipment: 20-06-2024
to see stock.
Add to basket

Description

Description

Accessories for filter holders for sample preparation.

AccessoriesXX4204708 Vent/Relief ValveYY3009053 O-rings, 90 mm filter housing, PTFEYY3009054 Support Screen, PTFE-coatedYY2209058 Underdrain Support, 90 mmYY2209059 Support Legs
Specifications

Specifications

Not Autoclavable
90 mm filter holders
Non-sterile
Hand Knob
Glass-filled Polypropylene, Brass
Hand Knob
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers