Learn More
Thermo Scientific™ Temperature Control Connectivity Accessories
Product Code. 11660991 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
1.8 m Interface Cable
5 ft. Interface Cable
5 m Interface Cable
Communication Extension Board
Software
For Use With (Equipment):
Cold/Heated Bath Circulators
RS-232 Communications Interface
Recirculating Chiller or Bath
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type For Use With (Equipment) unitSize
11660991 5 ft. Interface Cable Cold/Heated Bath Circulators Each
11608048 Software RS-232 Communications Interface Each
11650991 1.8 m Interface Cable Cold/Heated Bath Circulators Each
10252278 5 m Interface Cable Cold/Heated Bath Circulators Each
10765689 Communication Extension Board Recirculating Chiller or Bath Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11660991 Supplier Thermo Scientific™ Supplier No. 1600034
Maximize the efficiency of your temperature control device with Thermo Scientific™ Temperature Control Connectivity and monitoring accessories.
Connect your Thermo Scientific temperature control solution with your application.
  • Available for temperature control devices equipped with communications
  • Designed to work with your Thermo Scientific recirculating chiller or bath equipped with communications
  • Available in popular configurations that match your device’s connections and interface with your process
  • Take the guess work out of finding parts to match your recirculating chiller or bath
Thermo Scientific™ NEScom™ Windows-based software
  • Provides remote control of Thermo Scientific units equipped with an RS-232 communications interface
  • Choose upper and lower temperature alarm limits and monitor system status with an alarm
  • Record your results on a user selectable graph
  • Select from easy–to–use product icons
  • Create graphs and charts
  • Easily configure ramping functions over time
  • View pop-up alarm windows which display the error condition
  • View a virtual controller screen
  • PC compatible only
TRUSTED_SUSTAINABILITY
Specifications
Communications RS232, RS485
For Use With (Equipment) Cold/Heated Bath Circulators
Type 5 ft. Interface Cable