missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Accessories for Stainless Steel HPLC Columns

Product Code. 10082691 Shop All Macherey-Nagel Products
Change view
Click to view available options
Type:
Connector
Cutter Cone Fits
Locking Screw
Terminal Screw
Unit Size:
Each
Pack of 4
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
10082691 Locking Screw Pack of 4
10543412 Terminal Screw Pack of 5
10085711 Cutter Cone Fits Pack of 5
10072981 Connector Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10082691 Supplier Macherey-Nagel™ Supplier No. 718582
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Macherey-Nagel™ Accessories for Stainless Steel HPLC Columns include: plastic column end caps, capillary unions (100mm x 1/16 in. x 0.25mm), 1/16 in. nuts for connecting capillaries, and 1/16 in. ferrules.

Stainless steel columns are most frequently used in HPLC. The material is corrosion resistant, pressure stable and easy to work mechanically.

Specifications

Type Locking Screw
Color White
For Use With (Application) Locking Screw
Packaging Quantity 4 Pack
For Use With Analytical HPLC Columns
Material Plastic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.