missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Accessories for Vivapore™ Model Concentrators

Product Code. 10160801 Shop All Sartorius Products
Click to view available options
Type:
Expansion Reservoir
Recovery Pipet
Quantity:
10 Pack
100 Pack
For Use With (Equipment):
Sartorius Vivapore 10, 20 Concentrators
Sartorius Vivapore 5 Concentrator
Unit Size:
Pack of 10
Pack of 100
This item is not returnable. View return policy

Product Code. 10160801

Brand: Sartorius VPA006

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Items for various Vivapore models.

0
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Sartorius Vivapore 10, 20 Concentrators
Sample Volume 3 to 20 mL
Sterility Non-sterile
Quantity 10 Pack
Type Expansion Reservoir
Volume (Metric) 10 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.